Learn More
Medchemexpress LLC Ruthenium red (ammoniated ruthenium oxychloride) | 11103-72-3 | 99.95% | 786.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY103311
Encompass_Preferred
Ruthenium red, also known as ammoniated ruthenium oxychloride, is a polycationic dye primarily utilized for electron microscopy of biological samples. It strongly interacts with phospholipids, fatty acids, and acidic mucopolysaccharides. This compound also functions as an L-type calcium current (ICa) blocker.
  • Used for electron microscopy of cells, tissues, and vegetative bacteria.
  • Strongly reacts with phospholipids and fatty acids.
  • Binds to acidic mucopolysaccharides.
  • Acts as an L-type calcium current (ICa) blocker.
  • Blocks sarcoplasmic calcium release channels or mitochondrial calcium uptake.
  • Blocks sodium current and slows its inactivation time-course.
Catalog No. 50-003-64976