Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium Red, Technical Grade, Sigma-Aldrich

Catalog No. 01258821 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-821 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-821 Supplier MilliporeSigma Supelco Supplier No. R27511G
Add to Cart
Add to Cart

Ruthenium Red is a hexavalent polysaccharide stain. It inhibits mitochondrial Ca2+ uniporter. It is a capsaicin antagonist and blocks vanilloid receptor-1 (VR-1)-coupled ion channels.

TRUSTED_SUSTAINABILITY

Specifications

CAS 11103-72-3
Color Red
Quantity 1 g
Molecular Weight (g/mol) 786.35 g/mol
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.