Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ruthenium Red tetrahydrate, pure

Catalog No. AC191730050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191730050 5 g Glass bottle
AC191730010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191730050 Supplier Thermo Scientific Chemicals Supplier No. 191730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 12790-48-6 | Cl6H36N14O6Ru3-14 | 844.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12790-48-6
Molecular Formula Cl6H36N14O6Ru3-14
Molecular Weight (g/mol) 844.29
MDL Number MFCD00167176
InChI Key IMVKVBRYNZXYAP-UHFFFAOYSA-H
PubChem CID 108195
IUPAC Name azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate
SMILES [NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]

Specifications

Lambda Max 530 to 536nm
Molar Absorption Coefficient 8000L/(mole.cm) (lambda min., 0.02g/L H2O)
Packaging Glass bottle
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6·4H2O
Merck Index 10, 8161
Color Purple/Red
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonia
Quantity 5 g
Physical Form Crystalline Powder
Formula Weight 858.35
Percent Purity 34.6 to 36.1% (ruthenium)
Grade Pure
Chemical Name or Material Ruthenium red tetrahydrate, Pure
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.