Learn More
Polysciences, Inc. Ruthenium tetroxide, 0.5% stabilized aqueous solution
SDP
Catalog No. NC1929748
Encompass_Preferred
Ready to use RuO4solution is useful as a staining agent for electron microscopy of polymers, as well as a fixative for biological samples. Ruthenium tetroxide penetrates tissue very slowly, reacting strongly with proteins, glycogen and monosaccharides.
Catalog No. NC1929748 Supplier Polysciences, Inc. Supplier No. 1825325
Ready to use RuO4solution is useful as a staining agent for electron microscopy of polymers, as well as a fixative for biological samples. Ruthenium tetroxide penetrates tissue very slowly, reacting strongly with proteins, glycogen and monosaccharides.