Learn More
Apexbio Technology LLC Rutin 153-18-4 10g
SDP
Catalog No. 5000063296
Encompass_Preferred
Rutin (CAS 153-18-4) is a naturally occurring flavonoid glycoside that can be isolated from Sophora japonica L It exhibits antioxidant and anti-inflammatory activities primarily by scavenging reactive oxygen species modulating signaling pathways related to oxidative stress and inhibiting pro-inflammatory cytokine production Due to its pharmacological profile rutin is utilized extensively in biomedical research to investigate protective effects against cardiovascular dysfunction vascular permeability and inflammation-related conditions It is soluble in heated water methanol ethanol pyridine and alkaline environments and should be stored under cool dry and sealed conditions
Catalog No. 50-000-63296 Supplier Apexbio Technology LLC Supplier No. N183310000
Rutin (CAS 153-18-4) is a naturally occurring flavonoid glycoside that can be isolated from Sophora japonica L It exhibits antioxidant and anti-inflammatory activities primarily by scavenging reactive oxygen species modulating signaling pathways related to oxidative stress and inhibiting pro-inflammatory cytokine production Due to its pharmacological profile rutin is utilized extensively in biomedical research to investigate protective effects against cardiovascular dysfunction vascular permeability and inflammation-related conditions It is soluble in heated water methanol ethanol pyridine and alkaline environments and should be stored under cool dry and sealed conditions