Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutin, 97+%

Catalog No. AC132391000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132391000 100 g Plastic bottle
AC132390050 5 g Glass Bottle
AC132395000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC132391000 Supplier Thermo Scientific Chemicals Supplier No. 132391000
Only null left
Add to Cart

CAS: 207671-50-9 | C27H30O16 | 610.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207671-50-9
Molecular Formula C27H30O16
Molecular Weight (g/mol) 610.52
MDL Number MFCD01319140
InChI Key IKGXIBQEEMLURG-NVPNHPEKSA-N
Synonym rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid
PubChem CID 5280805
ChEBI CHEBI:28527
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 195.0°C
Color White
Loss on Drying 5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 8438
Solubility Information Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene
Formula Weight 664.58
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Rutin hydrate, 97.0-102.0%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
EINECSNumber 205-814-1
RTECSNumber VM2975000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.