Learn More
Medchemexpress LLC Rutin (Rutoside) | 153-18-4 | 610.52 | 500 MG
SDP
Catalog No. 5000365706
Encompass_Preferred
Rutin (Rutoside) is a flavonoid found in many plants. It exhibits a wide range of biological activities including anti-inflammatory, antidiabetic, antioxidant, neuroprotective, nephroprotective, hepatoprotective, and reducing Aβ oligomer activities. It is also a CBR1 inhibitor and can cross the blood-brain barrier. Rutin attenuates vancomycin-induced renal tubular cell apoptosis by suppressing apoptosis, mitochondrial dysfunction, and oxidative stress.
  • Anti-inflammatory activity
  • Antidiabetic activity
  • Antioxidant activity
  • Neuroprotective properties
  • Nephroprotective properties
  • Hepatoprotective properties
  • Reduces Aβ oligomer activities
  • CBR1 inhibitor
  • Can cross the blood-brain barrier
  • Attenuates vancomycin-induced renal tubular cell apoptosis
Catalog No. 50-003-65706 Supplier Medchemexpress LLC Supplier No. 500MGHYN0148
Rutin (Rutoside) is a flavonoid found in many plants. It exhibits a wide range of biological activities including anti-inflammatory, antidiabetic, antioxidant, neuroprotective, nephroprotective, hepatoprotective, and reducing Aβ oligomer activities. It is also a CBR1 inhibitor and can cross the blood-brain barrier. Rutin attenuates vancomycin-induced renal tubular cell apoptosis by suppressing apoptosis, mitochondrial dysfunction, and oxidative stress.
  • Anti-inflammatory activity
  • Antidiabetic activity
  • Antioxidant activity
  • Neuroprotective properties
  • Nephroprotective properties
  • Hepatoprotective properties
  • Reduces Aβ oligomer activities
  • CBR1 inhibitor
  • Can cross the blood-brain barrier
  • Attenuates vancomycin-induced renal tubular cell apoptosis