Learn More
AdipoGen Rutin trihydrate
SDP
Catalog No. 502562547
Encompass_Preferred
Chemical. CAS 250249-75-3. Formula C27H30O16 . 3H20. MW 610.5 . 54.0. Isolated from Ruta graveolens. Antioxidant. Free radical scavenger. Nitric oxide NO scavenger. Anticancer compound. Antiproliferative. Apoptosis inducer. Reduces induced DNA damage. Protective against carcinogenesis. Hypolipidaemic. Reduces triacylglycerol levels. Anti-inflammatory. Platelet aggregation inhibitor. Anti-hyperglycaemic. Anti-adipogenic. Insulin-mimetic. Activates synthesis of glucose transporter GLUT4. Neuroprotective. Cardioprotective. Reduces lipid peroxidation. Weak alpha-glucosidase inhibitor. Vasodilatory compound.
Catalog No. 50-256-2547 Supplier AdipoGen Supplier No. AGCN20408G005
Chemical. CAS 250249-75-3. Formula C27H30O16 . 3H20. MW 610.5 . 54.0. Isolated from Ruta graveolens. Antioxidant. Free radical scavenger. Nitric oxide NO scavenger. Anticancer compound. Antiproliferative. Apoptosis inducer. Reduces induced DNA damage. Protective against carcinogenesis. Hypolipidaemic. Reduces triacylglycerol levels. Anti-inflammatory. Platelet aggregation inhibitor. Anti-hyperglycaemic. Anti-adipogenic. Insulin-mimetic. Activates synthesis of glucose transporter GLUT4. Neuroprotective. Cardioprotective. Reduces lipid peroxidation. Weak alpha-glucosidase inhibitor. Vasodilatory compound.