Learn More
Medchemexpress LLC Rutin (trihydrate) | 250249-75-3 | 96.15% | 664.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW013075100MG
Encompass_Preferred
Rutin trihydrate is a multifunctional natural flavonoid glycoside that has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties, along with cardioprotective and neuroprotective activities. Rutin and/or its metabolites can cross the blood-brain barrier, modifying cognitive and various behavioral symptoms of neurodegenerative diseases.
  • Decreases acrylamide-induced cytotoxicity in PC12 cells.
  • Increases cell viability in PC12 cells.
  • Exhibits cardioprotective and neuroprotective activities.
  • Suitable for laboratory research.
  • Can be stored at 4°C, sealed, away from moisture and light.
Catalog No. 50-004-54352