Learn More
Medchemexpress LLC Rutin trihydrate | 250249-75-3 | ≥97.0% | 664.56 | 1 ML
SDP
Catalog No. 5000454183
Encompass_Preferred
Rutin trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties, as well as cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside.
  • Excellent antioxidant properties.
  • Anti-inflammatory properties.
  • Anti-diabetic properties.
  • Anti-carcinogenic properties.
  • Cardioprotective activities.
  • Neuroprotective activities.
  • Available in solution and solid forms.
  • Purity ≥97.0%.
Catalog No. 50-004-54183 Supplier Medchemexpress LLC Supplier No. HYW01307510MM/1ML
Rutin trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties, as well as cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside.
  • Excellent antioxidant properties.
  • Anti-inflammatory properties.
  • Anti-diabetic properties.
  • Anti-carcinogenic properties.
  • Cardioprotective activities.
  • Neuroprotective activities.
  • Available in solution and solid forms.
  • Purity ≥97.0%.