Learn More
Medchemexpress LLC Rutin (trihydrate) | 250249-75-3 | ≥97.0% | 664.56 | 10 G
SDP
Catalog No. 5000454575
Encompass_Preferred
Rutin (Rutoside) trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties, along with cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside.
  • Demonstrates excellent antioxidant properties.
  • Demonstrates excellent anti-inflammatory properties.
  • Demonstrates excellent anti-diabetic properties.
  • Demonstrates excellent anti-carcinogenic properties.
  • Exhibits cardioprotective activities.
  • Exhibits neuroprotective activities.
  • Decreases acrylamide (ACR)-induced cytotoxicity in PC12 cells.
  • May modify cognitive and behavioral symptoms of neurodegenerative diseases by crossing the blood-brain barrier.
Catalog No. 50-004-54575 Supplier Medchemexpress LLC Supplier No. HYW01307510G
Rutin (Rutoside) trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties, along with cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside.
  • Demonstrates excellent antioxidant properties.
  • Demonstrates excellent anti-inflammatory properties.
  • Demonstrates excellent anti-diabetic properties.
  • Demonstrates excellent anti-carcinogenic properties.
  • Exhibits cardioprotective activities.
  • Exhibits neuroprotective activities.
  • Decreases acrylamide (ACR)-induced cytotoxicity in PC12 cells.
  • May modify cognitive and behavioral symptoms of neurodegenerative diseases by crossing the blood-brain barrier.