Learn More
Medchemexpress LLC Rutin trihydrate | 250249-75-3 | ≥97.0% | 664.56 | 5 G
SDP
Catalog No. 5000454245
Encompass_Preferred
Rutin trihydrate is a multifunctional natural flavonoid glycoside with antioxidant, anti-inflammatory, anti-diabetic, anti-carcinogenic, cardioprotective, and neuroprotective properties. It has been shown to decrease acrylamide-induced cytotoxicity in PC12 cells and increase cell viability. Its ability to cross the blood-brain barrier allows it to modify cognitive and behavioral symptoms of neurodegenerative diseases. In vivo studies also show it causes behavioral and neurochemical changes.
  • Multifunctional natural flavonoid glycoside
  • Exhibits excellent antioxidant properties
  • Provides anti-inflammatory and anti-diabetic effects
  • Demonstrates anti-carcinogenic activity
  • Offers cardioprotective and neuroprotective activities
  • Decreases acrylamide-induced cytotoxicity in PC12 cells
  • Increases cell viability
  • Modifies cognitive and behavioral symptoms of neurodegenerative diseases
Catalog No. 50-004-54245 Supplier Medchemexpress LLC Supplier No. HYW0130755G
Rutin trihydrate is a multifunctional natural flavonoid glycoside with antioxidant, anti-inflammatory, anti-diabetic, anti-carcinogenic, cardioprotective, and neuroprotective properties. It has been shown to decrease acrylamide-induced cytotoxicity in PC12 cells and increase cell viability. Its ability to cross the blood-brain barrier allows it to modify cognitive and behavioral symptoms of neurodegenerative diseases. In vivo studies also show it causes behavioral and neurochemical changes.
  • Multifunctional natural flavonoid glycoside
  • Exhibits excellent antioxidant properties
  • Provides anti-inflammatory and anti-diabetic effects
  • Demonstrates anti-carcinogenic activity
  • Offers cardioprotective and neuroprotective activities
  • Decreases acrylamide-induced cytotoxicity in PC12 cells
  • Increases cell viability
  • Modifies cognitive and behavioral symptoms of neurodegenerative diseases