Learn More
Medchemexpress LLC Rutin trihydrate | 250249-75-3 | ≥97.0% | 664.56 | 50 MG
SDP
Catalog No. 5000454244
Encompass_Preferred
Rutin (Rutoside) trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties. It also exhibits cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside
  • Excellent antioxidant properties
  • Anti-inflammatory properties
  • Anti-diabetic properties
  • Anti-carcinogenic properties
  • Cardioprotective activities
  • Neuroprotective activities
Catalog No. 50-004-54244 Supplier Medchemexpress LLC Supplier No. HYW01307550MG
Rutin (Rutoside) trihydrate is a multifunctional natural flavonoid glycoside. It has demonstrated excellent antioxidant, anti-inflammatory, anti-diabetic, and anti-carcinogenic properties. It also exhibits cardioprotective and neuroprotective activities.
  • Multifunctional natural flavonoid glycoside
  • Excellent antioxidant properties
  • Anti-inflammatory properties
  • Anti-diabetic properties
  • Anti-carcinogenic properties
  • Cardioprotective activities
  • Neuroprotective activities