Learn More
AdipoGen Rutin trihydrate
SDP
Catalog No. 502560112
Encompass_Preferred
Chemical. CAS 250249-75-3. Formula C27H30O16 . 3H2O. MW 664.5. Synthetic. A polyphenolic flavonoid that acts as an antioxidant and NO scavenger. It can attenuate peroxide production in glial cells by acting as a free radical scavenger and protect renal cells from oxidative injury. Inclusion of rutin in the diet of rats significantly reduced the appearance of single-strand breaks in nuclear DNA caused by hepatocarcinogens aflatoxin B1 and N-nitrosodiumethylamine. This protection from DNA damage was found to be due to a reduction in the induction of repair enzymes polymerase, DNA polymerase beta and DNA ligase. Since DNA damage and inefficient repair are thought to initiate the process of carcinogenesis, effects of rutin on these functions suggests a protective role of this flavonoid against carcinogenesis induced by chemical carcinogens. Rutin ameliorates obesity through brown fat activation.
Catalog No. 50-256-0112 Supplier AdipoGen Supplier No. CDXR0066G500
Chemical. CAS 250249-75-3. Formula C27H30O16 . 3H2O. MW 664.5. Synthetic. A polyphenolic flavonoid that acts as an antioxidant and NO scavenger. It can attenuate peroxide production in glial cells by acting as a free radical scavenger and protect renal cells from oxidative injury. Inclusion of rutin in the diet of rats significantly reduced the appearance of single-strand breaks in nuclear DNA caused by hepatocarcinogens aflatoxin B1 and N-nitrosodiumethylamine. This protection from DNA damage was found to be due to a reduction in the induction of repair enzymes polymerase, DNA polymerase beta and DNA ligase. Since DNA damage and inefficient repair are thought to initiate the process of carcinogenesis, effects of rutin on these functions suggests a protective role of this flavonoid against carcinogenesis induced by chemical carcinogens. Rutin ameliorates obesity through brown fat activation.