Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutin trihydrate, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013705 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3705 25 mg
11-101-3704 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3705 Supplier MilliporeSigma Supelco Supplier No. 7809525MGF
Add to Cart
Add to Cart

Rutin trihydrate is a bioflavonoid, which may play a key role in the functioning of capillaries.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 250249-75-3
Physical Form Neat
Linear Formula C27H30O16 · 3H2 O
MDL Number MFCD00149490
Synonym Quercetin-3-rutinoside trihydrate; Vitamin P trihydrate
Type Analytical Standard
Quantity 25 mg
Formula Weight 664.56
Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.