Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutinose Heptaacetate, TCI America™
SDP

Catalog No. H06281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06281G Supplier TCI America Supplier No. H06281G

Chemical Identifiers

CAS 29202-64-0
Molecular Formula C26H36O17
MDL Number MFCD00161474
Synonym Hepta-O-acetyl Rutinose

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 1 g
Formula Weight 620.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Rutinose Heptaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.