Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutinose 95.0+%, TCI America™
SDP

Catalog No. R0062100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0062100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0062100MG Supplier TCI America Supplier No. R0062100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula C12H22O10
MDL Number MFCD00161473

Specifications

Color White-Yellow
CAS 90-74-4
Quantity 100 mg
Formula Weight 326.3
Percent Purity >95.0% (HPLC)
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.