Learn More
Medchemexpress LLC RuvB E.coli His-SU 50ug
SDP
Catalog No. 5000196692
Encompass_Preferred
Recombinant RuvB protein from Escherichia coli, expressed with an N-terminal 6xHis-SUMO tag and purified to greater than 90% purity. The full-length protein (336 amino acids; ~53.2 kDa) is supplied at research-scale quantity (50 μg) for biochemical, enzymatic, and structural studies of DNA repair and recombination.
  • Full-length protein (336 amino acids) for native activity
  • N-terminal His-SUMO tag aids purification and solubility
  • Purified to >90% as determined by reducing SDS-PAGE
  • Suitable for biochemical and structural assays of helicase activity
  • Supplied at research-scale quantity (50 μg) for assay development
Catalog No. 50-001-96692 Supplier Medchemexpress LLC Supplier No. HYP7151350UG
Recombinant RuvB protein from Escherichia coli, expressed with an N-terminal 6xHis-SUMO tag and purified to greater than 90% purity. The full-length protein (336 amino acids; ~53.2 kDa) is supplied at research-scale quantity (50 μg) for biochemical, enzymatic, and structural studies of DNA repair and recombination.
  • Full-length protein (336 amino acids) for native activity
  • N-terminal His-SUMO tag aids purification and solubility
  • Purified to >90% as determined by reducing SDS-PAGE
  • Suitable for biochemical and structural assays of helicase activity
  • Supplied at research-scale quantity (50 μg) for assay development