Learn More
Medchemexpress LLC RuvB E.coli His-SU 5ug
SDP
Catalog No. 5000195798
Encompass_Preferred
RuvB Protein, E. coli (His-SUMO) is a recombinant, full-length Holliday junction ATP-dependent DNA helicase expressed in Escherichia coli and supplied with an N-terminal 6xHis-SUMO affinity tag. Supplied as a purified protein in small-scale formats (for example, 5 μg), it is intended for biochemical assays, structural studies, and protein-protein interaction analyses that benefit from tag-enabled purification and detection.
  • Full-length sequence corresponding to residues M1-P336.
  • N-terminal 6xHis-SUMO tag for efficient affinity purification.
  • Expression in Escherichia coli for rapid bacterial production.
  • Purified protein supplied in small, assay-friendly quantities.
  • Suitable for biochemical assays, helicase activity testing, and interaction studies.
  • Molecular mass approx. 37,174 Da based on sequence.
Catalog No. 50-001-95798 Supplier Medchemexpress LLC Supplier No. HYP715135UG
RuvB Protein, E. coli (His-SUMO) is a recombinant, full-length Holliday junction ATP-dependent DNA helicase expressed in Escherichia coli and supplied with an N-terminal 6xHis-SUMO affinity tag. Supplied as a purified protein in small-scale formats (for example, 5 μg), it is intended for biochemical assays, structural studies, and protein-protein interaction analyses that benefit from tag-enabled purification and detection.
  • Full-length sequence corresponding to residues M1-P336.
  • N-terminal 6xHis-SUMO tag for efficient affinity purification.
  • Expression in Escherichia coli for rapid bacterial production.
  • Purified protein supplied in small, assay-friendly quantities.
  • Suitable for biochemical assays, helicase activity testing, and interaction studies.
  • Molecular mass approx. 37,174 Da based on sequence.