Learn More
Aviva Systems Biology Corp RUVBL1 Antibody - N-terminal region (ARP32349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RUVBL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RUVBL1. Alternate names: ECP54, NMP238, Pontin52, RVB1, TIH1, TIP49, TIP49A, INO80H, PONTINApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54539