Learn More
Aviva Systems Biology Corp RUVBL2 Antibody - N-terminal region (ARP32378_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32378_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RUVBL2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RUVBL2. Alternate names: RVB2, TIH2, ECP51, TIP48, CGI-46, INO80J, REPTIN, TIP49BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54553