Learn More
Medchemexpress LLC RuvC E.coli His-SU 10ug
SDP
Catalog No. 5000195900
Encompass_Preferred
Recombinant RuvC protein is an E. coli-expressed endonuclease that resolves Holliday junctions during homologous recombination and DNA repair. Supplied lyophilized with N-terminal SUMO and 6xHis affinity tags, it is formulated for biochemical and structural studies, with low endotoxin and high purity to support enzymatic assays.
  • E. coli expression for bacterial protein folding.
  • N-terminal SUMO and 6xHis tags for streamlined purification.
  • Greater than 90% purity as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Endotoxin <1 EU/μg to support sensitive assays.
  • Recommended aliquoting and storage at -20°C or -80°C to preserve activity.
Catalog No. 50-001-95900 Supplier Medchemexpress LLC Supplier No. HYP7151410UG
Recombinant RuvC protein is an E. coli-expressed endonuclease that resolves Holliday junctions during homologous recombination and DNA repair. Supplied lyophilized with N-terminal SUMO and 6xHis affinity tags, it is formulated for biochemical and structural studies, with low endotoxin and high purity to support enzymatic assays.
  • E. coli expression for bacterial protein folding.
  • N-terminal SUMO and 6xHis tags for streamlined purification.
  • Greater than 90% purity as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Endotoxin <1 EU/μg to support sensitive assays.
  • Recommended aliquoting and storage at -20°C or -80°C to preserve activity.