Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RuvC E.coli His-SU 50ug
SDP

Catalog No. 5000196601
Encompass_Preferred

RuvC Protein, E. coli (His-SUMO) is a lyophilized recombinant endonuclease derived from Escherichia coli that processes Holliday junctions during homologous recombination. The protein is supplied with an N-terminal SUMO and 6xHis tag, and is provided with quality data including purity and endotoxin testing to support molecular biology and biochemical applications.

  • Recombinant endonuclease expressed in E. coli with N-terminal SUMO and 6xHis tags.
  • Lyophilized powder formulation for long-term storage and transport.
  • Purity of at least 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in PBS with trehalose for stability after lyophilization.
  • Recommended reconstitution concentration of at least 100 μg/mL to preserve activity.
  • Stable storage options include -20°C (lyophilized) and -80°C for long-term aliquots.

Catalog No. 50-001-96601 Supplier Medchemexpress LLC Supplier No. HYP7151450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

RuvC Protein, E. coli (His-SUMO) is a lyophilized recombinant endonuclease derived from Escherichia coli that processes Holliday junctions during homologous recombination. The protein is supplied with an N-terminal SUMO and 6xHis tag, and is provided with quality data including purity and endotoxin testing to support molecular biology and biochemical applications.

  • Recombinant endonuclease expressed in E. coli with N-terminal SUMO and 6xHis tags.
  • Lyophilized powder formulation for long-term storage and transport.
  • Purity of at least 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in PBS with trehalose for stability after lyophilization.
  • Recommended reconstitution concentration of at least 100 μg/mL to preserve activity.
  • Stable storage options include -20°C (lyophilized) and -80°C for long-term aliquots.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.