Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RuvC E.coli His-SU 5ug
SDP

Catalog No. 5000196576
Encompass_Preferred

RuvC is a recombinant crossover junction endodeoxyribonuclease supplied as a His-SUMO fusion and expressed in Escherichia coli for use in biochemical and molecular biology assays. The product is supplied in small research sizes with documentation available from the manufacturer.

  • Recombinant endodeoxyribonuclease suitable for in vitro assays.
  • Expressed in E. coli with an N-terminal SUMO and 6xHis tag for ease of purification.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in small research sizes for assay development and optimization.
  • Certificate of analysis and safety data sheet available from the manufacturer.

Catalog No. 50-001-96576 Supplier Medchemexpress LLC Supplier No. HYP715145UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RuvC is a recombinant crossover junction endodeoxyribonuclease supplied as a His-SUMO fusion and expressed in Escherichia coli for use in biochemical and molecular biology assays. The product is supplied in small research sizes with documentation available from the manufacturer.

  • Recombinant endodeoxyribonuclease suitable for in vitro assays.
  • Expressed in E. coli with an N-terminal SUMO and 6xHis tag for ease of purification.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in small research sizes for assay development and optimization.
  • Certificate of analysis and safety data sheet available from the manufacturer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.