Learn More
Medchemexpress LLC HY-50856 10mg Medchemexpress, Ruxolitinib CAS:941678-49-5 Purity:>98%
SDP
Catalog No. 502029341
Encompass_Preferred
Medchemexpress, HY-50856 10mg Ruxolitinib CAS:941678-49-5 Ruxolitinib (INCB18424) is a potent and selective JAK1/2 inhibitor with IC50s of 3.3 nM and 2.8 nM in cell-free assays, and has 130-fold selectivity for JAK1/2 over JAK3. Ruxolitinib induces autophagy and kills tumor cells through toxic mitophagy[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9341 Supplier Medchemexpress LLC Supplier No. HY5085610MG
Medchemexpress, HY-50856 10mg Ruxolitinib CAS:941678-49-5 Ruxolitinib (INCB18424) is a potent and selective JAK1/2 inhibitor with IC50s of 3.3 nM and 2.8 nM in cell-free assays, and has 130-fold selectivity for JAK1/2 over JAK3. Ruxolitinib induces autophagy and kills tumor cells through toxic mitophagy[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.