Learn More
Medchemexpress LLC Ruxolitinib 500mg
SDP
Supplier:  Medchemexpress LLC HY50856500MG
Encompass_Preferred
Ruxolitinib (INCB18424) is a potent and selective JAK1/2 inhibitor with IC50s of 3.3 nM and 2.8 nM in cell-free assays, and has 130-fold selectivity for JAK1/2 over JAK3. Ruxolitinib induces autophagy and kills tumor cells through toxic mitophagy.
Catalog No. NC2152424