Learn More
Medchemexpress LLC HY-50856 50mg , Ruxolitinib CAS:941678-49-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5085650MG
Encompass_Preferred
HY-50856 50mg Ruxolitinib CAS: 941678-49-5 Ruxolitinib (INCB18424) is a potent and selective JAK1/2 inhibitor with IC50s of 3.3 nM and 2.8 nM in cell-free assays, and has 130-fold selectivity for JAK1/2 over JAK3. Ruxolitinib induces autophagy and kills tumor cells through toxic mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9114