Learn More
AdipoGen Ruxolitinib free base
SDP
Supplier:  AdipoGen AGCR13624M100
Encompass_Preferred
Chemical. CAS 941678-49-5. Formula C17H18N6. MW 306.4. Antineoplastic, anti-inflammatory and immunomodulating agent. Orally bioavailable potent ATP mimetic that inhibits both JAK1 and JAK2 with IC50 values of 2.7 and 4.5nM, respectively and is less selective for JAK3 IC50=322nM. Affects DC differentiation and function, leading to impaired T cell activation. Used in the treatment of myeloproliferative neoplasms and psoriasis. Anticancer agent. Shown to induce apoptosis and autophagy. Potent and selective inhibitor of HIV-1 replication and virus reactivation in vitro.
Catalog No. 50-256-2262