Learn More
AdipoGen Ruxolitinib phosphate salt
SDP
Catalog No. 502562254
Encompass_Preferred
Chemical. CAS 1092939-17-7. Formula C17H18N6 . H3O4P. MW 306.4 . 98.0. Water-soluble phosphate salt of Ruxolitinib. Antineoplastic, anti-inflammatory and immunomodulating agent. Orally bioavailable potent ATP mimetic that inhibits both JAK1 and JAK2 with IC50 values of 2.7 and 4.5nM, respectively and is less selective for JAK3 IC50=322nM. Affects DC differentiation and function, leading to impaired T cell activation. Used in the treatment of myeloproliferative neoplasms and psoriasis. Anticancer agent. Shown to induce apoptosis and autophagy. Potent and selective inhibitor of HIV-1 replication and virus reactivation in vitro.
Catalog No. 50-256-2254 Supplier AdipoGen Supplier No. AGCR13645M025
Chemical. CAS 1092939-17-7. Formula C17H18N6 . H3O4P. MW 306.4 . 98.0. Water-soluble phosphate salt of Ruxolitinib. Antineoplastic, anti-inflammatory and immunomodulating agent. Orally bioavailable potent ATP mimetic that inhibits both JAK1 and JAK2 with IC50 values of 2.7 and 4.5nM, respectively and is less selective for JAK3 IC50=322nM. Affects DC differentiation and function, leading to impaired T cell activation. Used in the treatment of myeloproliferative neoplasms and psoriasis. Anticancer agent. Shown to induce apoptosis and autophagy. Potent and selective inhibitor of HIV-1 replication and virus reactivation in vitro.