Learn More
TARGETMOL CHEMICALS INC RUXOLITINIB S ENANTIOMER 50MG
SDP
Catalog No. 502576588
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ruxolitinib S enantiomer (INCB18424) is the S-enantiomer of Ruxolitinib. Ruxolitinib is the first potent selective JAK1/2 inhibitor. purity: 99%
Catalog No. 50-257-6588 Supplier TARGETMOL CHEMICALS INC Supplier No. T615650MG
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ruxolitinib S enantiomer (INCB18424) is the S-enantiomer of Ruxolitinib. Ruxolitinib is the first potent selective JAK1/2 inhibitor. purity: 99%