Learn More
Medchemexpress LLC Ruxolitinib (S enantiomer) | 941685-37-6 | 99.6% | C17H18N6 | 100 MG
SDP
Catalog No. 5000358229
Encompass_Preferred
Ruxolitinib (S enantiomer) is the S-enantiomer of Ruxolitinib (HY-50856). It is an orally active, potent JAK inhibitor, intended for research use only.
  • Orally active
  • Potent JAK inhibitor
  • Suppresses IFNβ-induced expression of ISG, IFIT, and IFITM in human small airway epithelial cells (SAECs) at 10 μM for 12 hours
  • Reduces haematopoietic cell expansion in myeloproliferative neoplasms (MPN) in animal models (30 mg/kg, oral gavage, twice per day for 8 weeks)
Catalog No. 50-003-58229 Supplier Medchemexpress LLC Supplier No. HY50856A100MG
Ruxolitinib (S enantiomer) is the S-enantiomer of Ruxolitinib (HY-50856). It is an orally active, potent JAK inhibitor, intended for research use only.
  • Orally active
  • Potent JAK inhibitor
  • Suppresses IFNβ-induced expression of ISG, IFIT, and IFITM in human small airway epithelial cells (SAECs) at 10 μM for 12 hours
  • Reduces haematopoietic cell expansion in myeloproliferative neoplasms (MPN) in animal models (30 mg/kg, oral gavage, twice per day for 8 weeks)