Learn More
Medchemexpress LLC Ruxolitinib (S enantiomer) | 941685-37-6 | 99.6% | C17H18N6 | 50 MG
SDP
Catalog No. 5000358780
Encompass_Preferred
Ruxolitinib (S enantiomer), also known as S-Ruxolitinib, is the S-enantiomer of Ruxolitinib. It is an orally active and potent JAK inhibitor, widely utilized in research studies.
  • Orally active and potent JAK inhibitor
  • Suppresses IFNβ-induced gene expression in human small airway epithelial cells
  • Reduces haematopoietic cell expansion in myeloproliferative neoplasms in vivo
Catalog No. 50-003-58780 Supplier Medchemexpress LLC Supplier No. HY50856A50MG
Ruxolitinib (S enantiomer), also known as S-Ruxolitinib, is the S-enantiomer of Ruxolitinib. It is an orally active and potent JAK inhibitor, widely utilized in research studies.
  • Orally active and potent JAK inhibitor
  • Suppresses IFNβ-induced gene expression in human small airway epithelial cells
  • Reduces haematopoietic cell expansion in myeloproliferative neoplasms in vivo