Learn More
STEMCELL Technologies Ruxolitinib​, Size: 1 mg
SDP
Catalog No. 501976071
Encompass
Ruxolitinib is an ATP mimic that inhibits all Janus-associated kinase (JAK) family kinases with a preference for JAK-1 and JAK-2 over JAK-3 and TYK2 with IC₅₀ values of 3.3, 2.8, 428 and 19 nM respectively (Verstovsek; Quintás-Cardama et al.).CANCER RESEARCH· Suppresses erythroid progenitor colony formation from peripheral blood mononuclear cells of polycythemia vera patients with the constitutively active JAK2 V617F mutation. Reduces malignant cell proliferation and decreases interleukin-6 and TNF-α signaling in a JAK2 V617F-driven mouse model of myeloproliferative disorder (Quintás-Cardama et al.). OTHER· Promotes hair regrowth in a mouse model of alopecia areata (Xing et al.).
Catalog No. 50-197-6071 Supplier STEMCELL Technologies Supplier No. 73402
Ruxolitinib is an ATP mimic that inhibits all Janus-associated kinase (JAK) family kinases with a preference for JAK-1 and JAK-2 over JAK-3 and TYK2 with IC₅₀ values of 3.3, 2.8, 428 and 19 nM respectively (Verstovsek; Quintás-Cardama et al.).CANCER RESEARCH· Suppresses erythroid progenitor colony formation from peripheral blood mononuclear cells of polycythemia vera patients with the constitutively active JAK2 V617F mutation. Reduces malignant cell proliferation and decreases interleukin-6 and TNF-α signaling in a JAK2 V617F-driven mouse model of myeloproliferative disorder (Quintás-Cardama et al.). OTHER· Promotes hair regrowth in a mouse model of alopecia areata (Xing et al.).