Learn More
Medchemexpress LLC Ruxolitinib (Standard) | 941678-49-5 | 99.7% | C₁₇H₁₈N₆ | 50 MG
SDP
Catalog No. 5000357780
Encompass_Preferred
Ruxolitinib (Standard) is the analytical standard of Ruxolitinib, intended for research and analytical applications. It is a potent and selective JAK1/2 inhibitor, inducing autophagy and killing tumor cells through toxic mitophagy. This compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard in reference assays
  • Suitable for HPLC, GC, and MS research experiments
  • Store at 4°C, sealed, away from light and moisture
Catalog No. 50-003-57780 Supplier Medchemexpress LLC Supplier No. HY50856R50MG
Ruxolitinib (Standard) is the analytical standard of Ruxolitinib, intended for research and analytical applications. It is a potent and selective JAK1/2 inhibitor, inducing autophagy and killing tumor cells through toxic mitophagy. This compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard in reference assays
  • Suitable for HPLC, GC, and MS research experiments
  • Store at 4°C, sealed, away from light and moisture