Learn More
Selleck Chemical LLC Ruxotemitide
SDP
Catalog No. 5000000685
Encompass_Preferred
Ruxotemitide (LTX 315) is the oncolytic peptide that kills cancer cells through Bax/Bak-regulated mitochondrial membrane permeabilization
Catalog No. 50-000-00685 Supplier Selleck Chemical LLC Supplier No. S819925MG
Ruxotemitide (LTX 315) is the oncolytic peptide that kills cancer cells through Bax/Bak-regulated mitochondrial membrane permeabilization