Learn More
Selleck Chemical LLC Ruxotemitide5mg
SDP
Supplier:  Selleck Chemical LLC S81995MG
Encompass_Preferred
Ruxotemitide (LTX 315, Oncopore) is the oncolytic peptide that kills cancer cells through Bax/Bak-regulated mitochondrial membrane permeabilization. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3347