Learn More
MSC Classic Accessories 5th Wheel Hitch Cover, Heavy Duty Vinyl, Black
Catalog No. 502838311
Encompass
The Over Drive RV 5th Wheel Hitch Cover protects hitches from dirt, weather, and debris. This easy-to-attach cover offers reliable protection against various elements, ensuring the longevity of your hitch.
  • Three year limited warranty
  • Heavy duty water-resistant vinyl with a soft, non scratch backing
  • UV and mildew resistant
  • Protects against dirt, grease, rust, and harmful UV rays
  • Attaches with elastic cord and hook and or rip and grip straps
  • Fast and easy fit
Catalog No. 50-283-8311 Supplier MSC Supplier No. 36578854
The Over Drive RV 5th Wheel Hitch Cover protects hitches from dirt, weather, and debris. This easy-to-attach cover offers reliable protection against various elements, ensuring the longevity of your hitch.
  • Three year limited warranty
  • Heavy duty water-resistant vinyl with a soft, non scratch backing
  • UV and mildew resistant
  • Protects against dirt, grease, rust, and harmful UV rays
  • Attaches with elastic cord and hook and or rip and grip straps
  • Fast and easy fit