Learn More
Alkali Scientific RV Broth (USP) 10mL, 16x125mm Glass (5 Day QC), 10/Pk
SDP
Catalog No. 5000663220
Encompass_Preferred
RV Broth (USP) 10mL, 16x125mm Glass, 10/Pk is a selective enrichment medium designed for the isolation and detection of Salmonella species from food, water, and environmental samples. The formulation promotes preferential growth of Salmonella while suppressing competing microorganisms, improving detection sensitivity during microbiological testing. Each tube contains 10mL of broth in a 16x125mm screw cap glass tube that supports standardized incubation and sample handling procedures. The product undergoes extended quality control incubation to verify sterility and performance prior to release. Widely used in food safety laboratories, pharmaceutical quality control testing, and environmental microbiology workflows, this medium supports regulatory testing procedures requiring reliable enrichment of Salmonella organisms.
Catalog No. 50-006-63220 Supplier Alkali Scientific Supplier No. RVBT00
RV Broth (USP) 10mL, 16x125mm Glass, 10/Pk is a selective enrichment medium designed for the isolation and detection of Salmonella species from food, water, and environmental samples. The formulation promotes preferential growth of Salmonella while suppressing competing microorganisms, improving detection sensitivity during microbiological testing. Each tube contains 10mL of broth in a 16x125mm screw cap glass tube that supports standardized incubation and sample handling procedures. The product undergoes extended quality control incubation to verify sterility and performance prior to release. Widely used in food safety laboratories, pharmaceutical quality control testing, and environmental microbiology workflows, this medium supports regulatory testing procedures requiring reliable enrichment of Salmonella organisms.