Learn More
Grainger RV Extension 7 PK2
Catalog No. 502752284
Encompass_Preferred
The Haltec RV Extension is a 14 gauge, 25 foot long extension cord designed for RV and outdoor use. It has a 3 prong plug on one end and 3 socket receptacles on the other end, allowing you to plug in multiple devices. The cord is yellow for high visibility and has a lighted end for use in the dark. It is UL listed and rated for outdoor wet conditions. The 14 gauge wire allows it to handle up to 15 amps.
Catalog No. 50-275-2284 Supplier Grainger Supplier No. 141C48
The Haltec RV Extension is a 14 gauge, 25 foot long extension cord designed for RV and outdoor use. It has a 3 prong plug on one end and 3 socket receptacles on the other end, allowing you to plug in multiple devices. The cord is yellow for high visibility and has a lighted end for use in the dark. It is UL listed and rated for outdoor wet conditions. The 14 gauge wire allows it to handle up to 15 amps.