Learn More
Medchemexpress LLC RV01 | 1016897-10-1 | 98.1% | 263.29 g/mol | C17H13NO2 | 10MG
SDP
Catalog No. 5000235956
Encompass_Preferred
RV01 is a resveratrol analogue used as a research tool to probe ALDH2-related pathways and oxidative stress. It inhibits DNA damage, reduces ethanol-induced ALDH2 mRNA expression, scavenges hydroxyl radicals, and decreases iNOS expression, indicating anti-neuroinflammatory activity. Supplied as a powder for biochemical and cell-based research applications.
  • Inhibits DNA damage and reduces ALDH2 mRNA expression.
  • Scavenges hydroxyl radicals, indicating antioxidant activity.
  • Decreases iNOS expression and exhibits anti-neuroinflammatory effects.
  • Provided as a research-grade powder suitable for biochemical studies.
  • High purity for reliable experimental results.
Catalog No. 50-002-35956 Supplier Medchemexpress LLC Supplier No. HY12624110MG
RV01 is a resveratrol analogue used as a research tool to probe ALDH2-related pathways and oxidative stress. It inhibits DNA damage, reduces ethanol-induced ALDH2 mRNA expression, scavenges hydroxyl radicals, and decreases iNOS expression, indicating anti-neuroinflammatory activity. Supplied as a powder for biochemical and cell-based research applications.
  • Inhibits DNA damage and reduces ALDH2 mRNA expression.
  • Scavenges hydroxyl radicals, indicating antioxidant activity.
  • Decreases iNOS expression and exhibits anti-neuroinflammatory effects.
  • Provided as a research-grade powder suitable for biochemical studies.
  • High purity for reliable experimental results.