Learn More
Medchemexpress LLC Rv01 | 1016897-10-1 | 98.1% | 263.29 g/mol | C17H13NO2 | 50MG
SDP
Catalog No. 5000239614
Encompass_Preferred
RV01 is a resveratrol analogue that inhibits aldehyde dehydrogenase (ALDH) activity and reduces ethanol-induced ALDH2 mRNA expression. Supplied as a solid research compound for in vitro biochemical and cellular studies, it has documented solubility in DMSO and defined storage/stability conditions.
Catalog No. 50-002-39614 Supplier Medchemexpress LLC Supplier No. HY12624150MG
RV01 is a resveratrol analogue that inhibits aldehyde dehydrogenase (ALDH) activity and reduces ethanol-induced ALDH2 mRNA expression. Supplied as a solid research compound for in vitro biochemical and cellular studies, it has documented solubility in DMSO and defined storage/stability conditions.