Learn More
Medchemexpress LLC HY-126241 5mg Medchemexpress, RV01 CAS:1016897-10-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1262415MG
Encompass_Preferred
Medchemexpress, HY-126241 5mg RV01 CAS:1016897-10-1 RV01 is an analogue of resveratrol, inhibits DNA damage, reduces acetaldehyde dehydrogenase 2 (ALDH2) mRNA expression induced by ethanol, and exhibits hydroxyl radical scavenging activity. RV01 decreases iNOS expression, with anti-neuroinflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9721