Learn More
Electron Microscopy Sciences Edwards Rv3 Rotary Vacuum Pump, 220 V
SDP
Catalog No. 5032956
Encompass
Rotary vacuum pump with double stage. In high vacuum mode it is ideal for backing turbo pumps, analytical instruments, and electron microscopes. Configurable for high throughput mode. Ultra Grade 19 oil is recommended for use with the RV3. Speed - (Pneurop 6602). 50 Hz operation - 2 ft 3min-1 (3.3 m3h-1). 60 Hz operation - 2.3 ft 3min-1 (3.9 m 3h-1). Ultimate Vacuum (Total Pressure) - High Vacuum Mode - 2 x 10-3 mbar. Inlet connection - NW25.
Catalog No. 50-329-56 Supplier Electron Microscopy Sciences Supplier No. 91003E
Rotary vacuum pump with double stage. In high vacuum mode it is ideal for backing turbo pumps, analytical instruments, and electron microscopes. Configurable for high throughput mode. Ultra Grade 19 oil is recommended for use with the RV3. Speed - (Pneurop 6602). 50 Hz operation - 2 ft 3min-1 (3.3 m3h-1). 60 Hz operation - 2.3 ft 3min-1 (3.9 m 3h-1). Ultimate Vacuum (Total Pressure) - High Vacuum Mode - 2 x 10-3 mbar. Inlet connection - NW25.