Learn More
MARSHALL SCIENTIFIC Edwards RV5 Rotary Vane Vacuum Pump
SDP
Catalog No. NC2591257
Encompass_Preferred
The RV series of small oil sealed rotary vane pumps deliver excellent ultimate vacuum pressure, high pumping speeds and superior vapour handling capabilities with quiet operation. With over 300,000 units produced, these pumps offer proven performance that sets the industry standard for R&D and scientific pumping applications. All RV pumps are approved to UL and CSA standards by an external test house and feature our unique mode selector switch, meaning one model is suitable for both high vacuum or high throughput applications.
Catalog No. NC2591257 Supplier MARSHALL SCIENTIFIC Supplier No. EDA65301906
Add to Cart
Add to Cart
Request a Quote
The RV series of small oil sealed rotary vane pumps deliver excellent ultimate vacuum pressure, high pumping speeds and superior vapour handling capabilities with quiet operation. With over 300,000 units produced, these pumps offer proven performance that sets the industry standard for R&D and scientific pumping applications. All RV pumps are approved to UL and CSA standards by an external test house and feature our unique mode selector switch, meaning one model is suitable for both high vacuum or high throughput applications.