Learn More
Cayman Chemical RValerolacton 100mg
SDP
Catalog No. 502981944
Encompass_Preferred
An isomer of GVL, which is a prodrug to GHV; has potential for use as a biofuel
Catalog No. 50-298-1944 Supplier Cayman Chemical Supplier No. 20837100 MG
An isomer of GVL, which is a prodrug to GHV; has potential for use as a biofuel