Learn More
Cayman Chemical Rvaroxabn 10mg
SDP
Catalog No. 502970191
Encompass_Preferred
An orally active, direct inhibitor of Factor Xa (Ki = 0.4 nM) that inhibits thrombin formation without prolonging bleeding time; approved for clinical use as an anticoagulant in the prevention of stroke and the treatment of venous thromboembolisms
Catalog No. 50-297-0191 Supplier Cayman Chemical Supplier No. 1604310 MG
An orally active, direct inhibitor of Factor Xa (Ki = 0.4 nM) that inhibits thrombin formation without prolonging bleeding time; approved for clinical use as an anticoagulant in the prevention of stroke and the treatment of venous thromboembolisms