Learn More
Cayman Chemical Rvaroxabn 50mg
SDP
Catalog No. 502971861
Encompass_Preferred
An orally active, direct inhibitor of Factor Xa (Ki = 0.4 nM) that inhibits thrombin formation without prolonging bleeding time; approved for clinical use as an anticoagulant in the prevention of stroke and the treatment of venous thromboembolisms
Catalog No. 50-297-1861 Supplier Cayman Chemical Supplier No. 1604350 MG
An orally active, direct inhibitor of Factor Xa (Ki = 0.4 nM) that inhibits thrombin formation without prolonging bleeding time; approved for clinical use as an anticoagulant in the prevention of stroke and the treatment of venous thromboembolisms