Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RVD-Hpα TFA | 1431329-51-9 | 99.3% | 1538.67 | 5 MG
SDP

Catalog No. 5000424646
Encompass_Preferred

RVD-Hpα TFA is the N-terminally extended form of human hemopressin. It acts as a selective CB1 receptor agonist and increases intracellular Ca2+ levels in cells expressing CB1 receptors in vitro. It also functions as a high-affinity CB2 positive allosteric modulator (Ki=50 nM).

  • N-terminally extended form of human hemopressin.
  • Selective CB1 receptor agonist.
  • Increases intracellular Ca2+ levels in CB1 receptor expressing cells in vitro.
  • High-affinity CB2 positive allosteric modulator (Ki=50 nM).
  • Appears as a white to off-white solid.
  • Ships at room temperature in the continental US.
  • Store sealed and away from moisture.
  • Powder storage: -80°C for 2 years, -20°C for 1 year.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-24646 Supplier Medchemexpress LLC Supplier No. HYP1397A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RVD-Hpα TFA is the N-terminally extended form of human hemopressin. It acts as a selective CB1 receptor agonist and increases intracellular Ca2+ levels in cells expressing CB1 receptors in vitro. It also functions as a high-affinity CB2 positive allosteric modulator (Ki=50 nM).

  • N-terminally extended form of human hemopressin.
  • Selective CB1 receptor agonist.
  • Increases intracellular Ca2+ levels in CB1 receptor expressing cells in vitro.
  • High-affinity CB2 positive allosteric modulator (Ki=50 nM).
  • Appears as a white to off-white solid.
  • Ships at room temperature in the continental US.
  • Store sealed and away from moisture.
  • Powder storage: -80°C for 2 years, -20°C for 1 year.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.