Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RVD-Hpα TFA | 1431329-51-9 | 99.32% | 1538.67 | 10 MG
SDP

Catalog No. 5000422304
Encompass_Preferred

RVD-Hpα TFA is the N-terminally extended form of human hemopressin that acts as a selective CB1 receptor agonist. It increases intracellular Ca2+ levels in cells expressing CB1 receptors in vitro, and also functions as a high-affinity CB2 positive allosteric modulator (Ki=50 nM).

  • Molecular weight: 1538.67
  • Chemical formula: C67H106F3N19O19
  • Appearance: Solid
  • Color: White to off-white
  • Sequence shortening: RVDPVNFKLLSH
  • Related to peptide and derivative classifications, and acts as an inhibitor and substrate
  • Associated with blood or cardio-cerebrovascular disease and cardiovascular disease research
  • Involves GPCR/G protein and neuronal signaling pathways
  • Modulates cannabinoid receptors

Catalog No. 50-004-22304 Supplier Medchemexpress LLC Supplier No. HYP1397A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RVD-Hpα TFA is the N-terminally extended form of human hemopressin that acts as a selective CB1 receptor agonist. It increases intracellular Ca2+ levels in cells expressing CB1 receptors in vitro, and also functions as a high-affinity CB2 positive allosteric modulator (Ki=50 nM).

  • Molecular weight: 1538.67
  • Chemical formula: C67H106F3N19O19
  • Appearance: Solid
  • Color: White to off-white
  • Sequence shortening: RVDPVNFKLLSH
  • Related to peptide and derivative classifications, and acts as an inhibitor and substrate
  • Associated with blood or cardio-cerebrovascular disease and cardiovascular disease research
  • Involves GPCR/G protein and neuronal signaling pathways
  • Modulates cannabinoid receptors

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.