Learn More
Medchemexpress LLC RVG-Cys | 1186105-01-0 | 99.9% | 3369.77 | 5 MG
SDP
Catalog No. 5000396998
Encompass_Preferred
RVG-Cys (RVG29-Cys) is a peptide derived from rabies virus glycoprotein (RVG29) with Cys attached to facilitate subsequent conjugation. It enhances the specific targeted delivery of proteins in brain tissue and neurons.
  • Can be coupled to nano-carriers to enhance brain-targeted delivery and prolong accumulation time in the brain.
  • Promotes axonal regeneration and functional recovery in spinal cord injury models when incorporated into miR-NPs.
  • For research use only.
Catalog No. 50-003-96998 Supplier Medchemexpress LLC Supplier No. HYP5623A5MG
RVG-Cys (RVG29-Cys) is a peptide derived from rabies virus glycoprotein (RVG29) with Cys attached to facilitate subsequent conjugation. It enhances the specific targeted delivery of proteins in brain tissue and neurons.
  • Can be coupled to nano-carriers to enhance brain-targeted delivery and prolong accumulation time in the brain.
  • Promotes axonal regeneration and functional recovery in spinal cord injury models when incorporated into miR-NPs.
  • For research use only.